Anti-C4 binding protein B Mouse Monoclonal Antibody [clone: 1G9]

Supplier: Novus Biologicals
H00000725-M01
103322-906EA 798.46 USD
103322-906
Anti-C4 binding protein B Mouse Monoclonal Antibody [clone: 1G9]
Antibodies
The C4 binding protein B Antibody (1G9) from Novus Biologicals is a mouse monoclonal antibody to C4 binding protein B. This antibody reacts with human. The C4 binding protein B Antibody (1G9) has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation.

Type: Primary
Antigen: C4 binding protein B
Clonality: Monoclonal
Clone: 1G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR