Anti-Buster3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91707
103280-726EA 734.58 USD
103280-726
Anti-Buster3 Rabbit Polyclonal Antibody
Antibodies
The Buster3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Buster3. This antibody reacts with human. The Buster3 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Buster3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR