Anti-Blooms Syndrome Protein Blm Rabbit Polyclonal Antibody

NBP2-15594
102706-290EA 493.85 USD
102706-290
Anti-Blooms Syndrome Protein Blm Rabbit Polyclonal Antibody
The Blooms Syndrome Protein Blm Antibody from Novus Biologicals is a rabbit polyclonal antibody to Blooms Syndrome Protein Blm. This antibody reacts with human. The Blooms Syndrome Protein Blm Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Blooms Syndrome Protein Blm
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR