Anti-Blooms Syndrome Protein Blm Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-89929
103280-000EA 734.58 USD
103280-000
Anti-Blooms Syndrome Protein Blm Rabbit Polyclonal Antibody
Antibodies
The Blooms Syndrome Protein Blm Antibody from Novus Biologicals is a rabbit polyclonal antibody to Blooms Syndrome Protein Blm. This antibody reacts with human. The Blooms Syndrome Protein Blm Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Blooms Syndrome Protein Blm
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR