Anti-Birch pollen Bet V1 Human Monoclonal Antibody [clone: H.G3A3C2.04]

Supplier: Novus Biologicals
DDX0075P-100
103318-688EA 932.74 USD
103318-688
Anti-Birch pollen Bet V1 Human Monoclonal Antibody [clone: H.G3A3C2.04]
Antibodies
The Birch pollen Bet V1 Antibody (H.G3A3C2.04) from Novus Biologicals is a human monoclonal antibody to Birch pollen Bet V1. This antibody reacts with plant. The Birch pollen Bet V1 Antibody (H.G3A3C2.04) has been validated for the following applications: ELISA.

Type: Primary
Antigen: Birch pollen Bet V1
Clonality: Monoclonal
Clone: H.G3A3C2.04
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG2
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR