Anti-Birch pollen Bet V1 Human Monoclonal Antibody [clone: H.G28C10.01]

Supplier: Novus Biologicals
DDX0073P-100
103318-682EA 932.74 USD
103318-682
Anti-Birch pollen Bet V1 Human Monoclonal Antibody [clone: H.G28C10.01]
Antibodies
The Birch pollen Bet V1 Antibody (H.G28C10.01) from Novus Biologicals is a human monoclonal antibody to Birch pollen Bet V1. This antibody reacts with plant. The Birch pollen Bet V1 Antibody (H.G28C10.01) has been validated for the following applications: ELISA.

Type: Primary
Antigen: Birch pollen Bet V1
Clonality: Monoclonal
Clone: H.G28C10.01
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG4 Kappa
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR