Anti-Biotin (Vitamin B7 or Vitamin H) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: SPM375]

Supplier: Novus Biologicals
NBP2-34771PE
103405-150EA 638.77 USD
103405-150
Anti-Biotin (Vitamin B7 or Vitamin H) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: SPM375]
Antibodies
The Biotin (Vitamin B7 or Vitamin H) Antibody (SPM375) [PE] from Novus Biologicals is a mouse monoclonal antibody to Biotin (Vitamin B7 or Vitamin H). This antibody reacts with all species. The Biotin (Vitamin B7 or Vitamin H) Antibody (SPM375) [PE] has been validated for the following applications: Flow Cytometry.

Type: Primary
Antigen: Biotin (Vitamin B7 or Vitamin H)
Clonality: Monoclonal
Clone: SPM375
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR