Anti-Biotin (Vitamin B7 or Vitamin H) Mouse Monoclonal Antibody (DyLight 488) [clone: Hyb-8]

Supplier: Novus Biologicals
NBP2-34646G
103405-112EA 638.77 USD
103405-112
Anti-Biotin (Vitamin B7 or Vitamin H) Mouse Monoclonal Antibody (DyLight 488) [clone: Hyb-8]
Antibodies
The Biotin (Vitamin B7 or Vitamin H) Antibody (Hyb-8) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to Biotin (Vitamin B7 or Vitamin H). This antibody reacts with all species. The Biotin (Vitamin B7 or Vitamin H) Antibody (Hyb-8) [DyLight 488] has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: Biotin (Vitamin B7 or Vitamin H)
Clonality: Monoclonal
Clone: Hyb-8
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR