Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Bilirubin Oxidase Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-44156
The Bilirubin Oxidase Antibody from Novus Biologicals is a goat polyclonal antibody to Bilirubin Oxidase. This antibody reacts with fungi. The Bilirubin Oxidase Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation.

Type: Primary
Antigen: Bilirubin Oxidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Fungi




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
2 ml NBP2-44156 103408-286 Each (2ml) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Name Bilirubin Oxidase
Reactivity Fungi
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Goat
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Concentration 95 mg/ml
Immunogen Bilirubin Oxidase (Myrothecium verrucaria)
Purification Delipidation and Defibrination
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Storage Temperature Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty