Anti-beta-TrCP1/BTRC Mouse Monoclonal Antibody [clone: 2H2]

Supplier: Novus Biologicals
NBP2-46588
103405-100EA 663.82 USD
103405-100
Anti-beta-TrCP1/BTRC Mouse Monoclonal Antibody [clone: 2H2]
Antibodies
The beta-TrCP1 / BTRC Antibody (2H2) from Novus Biologicals is a mouse monoclonal antibody to beta-TrCP1 / BTRC. This antibody reacts with human. The beta-TrCP1 / BTRC Antibody (2H2) has been validated for the following applications: Western Blot, Flow Cytometry, Immunofluorescence.

Type: Primary
Antigen: beta-TrCP1/BTRC
Clonality: Monoclonal
Clone: 2H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR