Anti-beta-Synuclein Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-90342
103277-788EA 734.58 USD
103277-788
Anti-beta-Synuclein Rabbit Polyclonal Antibody
Antibodies
The beta-Synuclein Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta-Synuclein. This antibody reacts with human, mouse. The beta-Synuclein Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta-Synuclein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR