Anti-beta-Synuclein Mouse Monoclonal Antibody [clone: 3G6]

Supplier: Novus Biologicals
H00006620-M02
103310-434EA 798.46 USD
103310-434
Anti-beta-Synuclein Mouse Monoclonal Antibody [clone: 3G6]
Antibodies
The beta-Synuclein Antibody (3G6) from Novus Biologicals is a mouse monoclonal antibody to beta-Synuclein. This antibody reacts with human. The beta-Synuclein Antibody (3G6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: beta-Synuclein
Clonality: Monoclonal
Clone: 3G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR