Anti-beta-Synuclein Mouse Monoclonal Antibody [clone: 4F11]

Supplier: Novus Biologicals
H00006620-M16
103310-438EA 798.46 USD
103310-438
Anti-beta-Synuclein Mouse Monoclonal Antibody [clone: 4F11]
Antibodies
The beta-Synuclein Antibody (4F11) from Novus Biologicals is a mouse monoclonal antibody to beta-Synuclein. This antibody reacts with human. The beta-Synuclein Antibody (4F11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: beta-Synuclein
Clonality: Monoclonal
Clone: 4F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR