Anti-beta-Synuclein Mouse Monoclonal Antibody [clone: 3H4]

Supplier: Novus Biologicals
H00006620-M07
103310-436EA 798.46 USD
103310-436
Anti-beta-Synuclein Mouse Monoclonal Antibody [clone: 3H4]
Antibodies
The beta-Synuclein Antibody (3H4) from Novus Biologicals is a mouse monoclonal antibody to beta-Synuclein. This antibody reacts with human. The beta-Synuclein Antibody (3H4) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: beta-Synuclein
Clonality: Monoclonal
Clone: 3H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR