Anti-beta-NGF Mouse Monoclonal Antibody (DyLight 680) [clone: 25623]

Supplier: Novus Biologicals
NB120-10513FR
103405-088EA 493.97 USD
103405-088
Anti-beta-NGF Mouse Monoclonal Antibody (DyLight 680) [clone: 25623]
Antibodies
The beta-NGF Antibody (25623) [DyLight 680] from Novus Biologicals is a mouse monoclonal antibody to beta-NGF. This antibody reacts with human, mouse, rat. The beta-NGF Antibody (25623) [DyLight 680] has been validated for the following applications: ELISA.

Type: Primary
Antigen: beta-NGF
Clonality: Monoclonal
Clone: 25623
Conjugation: DyLight 680
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR