Anti-beta-NGF Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-19536
102709-944EA 635.51 USD
102709-944
Anti-beta-NGF Rabbit Polyclonal Antibody
Antibodies
The beta-NGF Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta-NGF. This antibody reacts with human. The beta-NGF Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: beta-NGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR