Anti-beta-III Tubulin Mouse Monoclonal Antibody (DyLight 350) [clone: TU-20]

Supplier: Novus Biologicals
NB600-1018UV
103405-050EA 706.28 USD
103405-050
Anti-beta-III Tubulin Mouse Monoclonal Antibody (DyLight 350) [clone: TU-20]
Antibodies
The beta-III Tubulin Antibody (TU-20) [DyLight 350] from Novus Biologicals is a mouse monoclonal antibody to beta-III Tubulin. This antibody reacts with human, mouse, all species. The beta-III Tubulin Antibody (TU-20) [DyLight 350] has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta-III Tubulin
Clonality: Monoclonal
Clone: TU-20
Conjugation: DyLight 350
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, All Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR