Anti-beta-Glucuronidase/GUSB Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00002990-B01P
103327-772EA 798.46 USD
103327-772
Anti-beta-Glucuronidase/GUSB Mouse Polyclonal Antibody
Antibodies
The beta-Glucuronidase / GUSB Antibody from Novus Biologicals is a mouse polyclonal antibody to beta-Glucuronidase / GUSB. This antibody reacts with human. The beta-Glucuronidase / GUSB Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: beta-Glucuronidase/GUSB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR