Anti-beta-Galactosidase-1/GLB1 Mouse Monoclonal Antibody [clone: 1C9]

Supplier: Novus Biologicals
NBP2-45731
103402-948EA 663.82 USD
103402-948
Anti-beta-Galactosidase-1/GLB1 Mouse Monoclonal Antibody [clone: 1C9]
Antibodies
The beta-Galactosidase-1 / GLB1 Antibody (1C9) from Novus Biologicals is a mouse monoclonal antibody to beta-Galactosidase-1 / GLB1. This antibody reacts with human, canine, monkey. The beta-Galactosidase-1 / GLB1 Antibody (1C9) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta-Galactosidase-1/GLB1
Clonality: Monoclonal
Clone: 1C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Canine, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR