Anti-beta-Catenin Mouse Monoclonal Antibody (Biotin) [clone: 12F7]

Supplier: Novus Biologicals
NBP1-54467B
103402-902EA 675.69 USD
103402-902
Anti-beta-Catenin Mouse Monoclonal Antibody (Biotin) [clone: 12F7]
Antibodies
The beta-Catenin Antibody (12F7) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to beta-Catenin. This antibody reacts with human, mouse, rat, chicken, primate. The beta-Catenin Antibody (12F7) [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta-Catenin
Clonality: Monoclonal
Clone: 12F7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chicken, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR