Anti-beta-Catenin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-89990
103277-412EA 805.35 USD
103277-412
Anti-beta-Catenin Rabbit Polyclonal Antibody
Antibodies
The beta-Catenin Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta-Catenin. This antibody reacts with human. The beta-Catenin Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta-Catenin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR