Anti-beta-Arrestin 1 Rabbit Polyclonal Antibody

H00000408-D01P
103322-164EA 627.69 USD
103322-164
Anti-beta-Arrestin 1 Rabbit Polyclonal Antibody
The beta-Arrestin 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta-Arrestin 1. This antibody reacts with human. The beta-Arrestin 1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: beta-Arrestin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR