Anti-beta-2 Adrenergic R/ADRB2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15564
102706-230EA 635.51 USD
102706-230
Anti-beta-2 Adrenergic R/ADRB2 Rabbit Polyclonal Antibody
Antibodies
The beta-2 Adrenergic R / ADRB2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta-2 Adrenergic R / ADRB2. This antibody reacts with human, rat. The beta-2 Adrenergic R / ADRB2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta-2 Adrenergic R/ADRB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR