Anti-beta-2 Adrenergic R/ADRB2 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00000154-B01P
103321-476EA 399 USD
103321-476
Anti-beta-2 Adrenergic R/ADRB2 Mouse Polyclonal Antibody
Antibodies
The beta-2 Adrenergic R / ADRB2 Antibody from Novus Biologicals is a mouse polyclonal antibody to beta-2 Adrenergic R / ADRB2. This antibody reacts with human. The beta-2 Adrenergic R / ADRB2 Antibody has been validated for the following applications: Western Blot, Flow Cytometry.

Type: Primary
Antigen: beta-2 Adrenergic R/ADRB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR