Anti-beta-1,3-Glucuronyltransferase 3/B3GAT3 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00026229-B01P
103342-204EA 798.46 USD
103342-204
Anti-beta-1,3-Glucuronyltransferase 3/B3GAT3 Mouse Polyclonal Antibody
Antibodies
The beta-1,3-Glucuronyltransferase 3 / B3GAT3 Antibody from Novus Biologicals is a mouse polyclonal antibody to beta-1,3-Glucuronyltransferase 3 / B3GAT3. This antibody reacts with human. The beta-1,3-Glucuronyltransferase 3 / B3GAT3 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: beta-1,3-Glucuronyltransferase 3/B3GAT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR