Anti-beta-1,3-Glucuronyltransferase 1/B3GAT1 Mouse Monoclonal Antibody (DyLight 350) [clone: HNK-1 or Leu-7]

NBP2-47787UV
103402-808EA 466.15 USD
103402-808
Anti-beta-1,3-Glucuronyltransferase 1/B3GAT1 Mouse Monoclonal Antibody (DyLight 350) [clone: HNK-1 or Leu-7]
The beta-1,3-Glucuronyltransferase 1 / B3GAT1 Antibody (HNK-1 or Leu-7) [DyLight 350] from Novus Biologicals is a mouse monoclonal antibody to beta-1,3-Glucuronyltransferase 1 / B3GAT1. This antibody reacts with human, rat (negative). The beta-1,3-Glucuronyltransferase 1 / B3GAT1 Antibody (HNK-1 or Leu-7) [DyLight 350] has been validated for the following applications: Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta-1,3-Glucuronyltransferase 1/B3GAT1
Clonality: Monoclonal
Clone: HNK-1 or Leu-7
Conjugation: DyLight 350
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human, Rat (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR