Anti-beta Tubulin Mouse Monoclonal Antibody (DyLight 550) [clone: BT7R]

Supplier: Novus Biologicals
NBP2-37830R
103404-988EA 706.28 USD
103404-988
Anti-beta Tubulin Mouse Monoclonal Antibody (DyLight 550) [clone: BT7R]
Antibodies
The beta Tubulin Antibody (BT7R) [DyLight 550] from Novus Biologicals is a mouse monoclonal antibody to beta Tubulin. This antibody reacts with human, mouse, rat, avian, chicken, primate, rabbit. The beta Tubulin Antibody (BT7R) [DyLight 550] has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: beta Tubulin
Clonality: Monoclonal
Clone: BT7R
Conjugation: DyLight 550
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Avian, Chicken, Primate, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR