Anti-beta Tubulin Mouse Monoclonal Antibody (Biotin) [clone: BT7R]

Supplier: Novus Biologicals
NBP2-37830B
103404-978EA 706.28 USD
103404-978
Anti-beta Tubulin Mouse Monoclonal Antibody (Biotin) [clone: BT7R]
Antibodies
The beta Tubulin Antibody (BT7R) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to beta Tubulin. This antibody reacts with human, mouse, rat, avian, chicken, primate, rabbit. The beta Tubulin Antibody (BT7R) [Biotin] has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: beta Tubulin
Clonality: Monoclonal
Clone: BT7R
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Avian, Chicken, Primate, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR