Anti-beta Tubulin Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1B12]

Supplier: Novus Biologicals
NBP2-50061F
103404-812EA 614.15 USD
103404-812
Anti-beta Tubulin Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1B12]
Antibodies
The beta Tubulin Antibody (1B12) [FITC] from Novus Biologicals is a mouse monoclonal antibody to beta Tubulin. This antibody reacts with human, mouse, rat, porcine, bovine. The beta Tubulin Antibody (1B12) [FITC] has been validated for the following applications: Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: beta Tubulin
Clonality: Monoclonal
Clone: 1B12
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat, Porcine, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR