Anti-beta Tubulin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15584
102706-270EA 635.51 USD
102706-270
Anti-beta Tubulin Rabbit Polyclonal Antibody
Antibodies
The beta Tubulin Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta Tubulin. This antibody reacts with human, mouse. The beta Tubulin Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta Tubulin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR