Anti-beta Tubulin Rabbit Polyclonal Antibody

NBP2-15583
102706-268EA 493.85 USD
102706-268
Anti-beta Tubulin Rabbit Polyclonal Antibody
The beta Tubulin Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta Tubulin. This antibody reacts with human, mouse. The beta Tubulin Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta Tubulin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR