Anti-beta Tubulin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56459SS
103359-172EA 208 USD
103359-172
Anti-beta Tubulin Rabbit Polyclonal Antibody
Antibodies
The beta Tubulin Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta Tubulin. This antibody reacts with human, mouse, rat, bovine, canine, chicken, feline, monkey, primate, xenopus. The beta Tubulin Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.

Simple Western,

Type: Primary
Antigen: beta Tubulin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Canine, Chicken, Feline, Monkey, Primate, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR