Anti-beta Tubulin Mouse Monoclonal Antibody [clone: 40000]

Supplier: Novus Biologicals
NBP2-50047
103404-822EA 491.08 USD
103404-822
Anti-beta Tubulin Mouse Monoclonal Antibody [clone: 40000]
Antibodies
The beta Tubulin Antibody (4E4) from Novus Biologicals is a mouse monoclonal antibody to beta Tubulin. This antibody reacts with human, mouse, rat. The beta Tubulin Antibody (4E4) has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: beta Tubulin
Clonality: Monoclonal
Clone: 40000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR