Anti-beta II Tubulin B Mouse Monoclonal Antibody (DyLight 350) [clone: 5B2]

Supplier: Novus Biologicals
NBP1-74037UV
103404-960EA 706.28 USD
103404-960
Anti-beta II Tubulin B Mouse Monoclonal Antibody (DyLight 350) [clone: 5B2]
Antibodies
The beta II Tubulin B Antibody (5B2) [DyLight 350] from Novus Biologicals is a mouse monoclonal antibody to beta II Tubulin B. This antibody reacts with human, mouse. The beta II Tubulin B Antibody (5B2) [DyLight 350] has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: beta II Tubulin B
Clonality: Monoclonal
Clone: 5B2
Conjugation: DyLight 350
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR