Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-beta II Tubulin B Mouse Monoclonal Antibody (DyLight 680) [clone: 5B3]

Supplier: Novus Biologicals
The beta II Tubulin B Antibody (5B3) [DyLight 680] from Novus Biologicals is a mouse monoclonal antibody to beta II Tubulin B. This antibody reacts with human, mouse. The beta II Tubulin B Antibody (5B3) [DyLight 680] has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: beta II Tubulin B
Clonality: Monoclonal
Clone: 5B3
Conjugation: DyLight 680
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-74029FR 103402-794 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Name beta II Tubulin B
Reactivity Human, Mouse
Conjugation DyLight® 680
Clonality Monoclonal
Antibody Type Primary
Clone 5B3
Host Mouse
Isotype IgG2a Kappa
Western Blot Yes
ImmunoFluorescence Yes
Antigen Synonyms tubulin beta-2B chain, class II beta-tubulin isotype, tubulin, beta polypeptide paralog, FLJ98847, MGC8685, bA506K6.1, DKFZp566F223, beta 2B
Gene ID 347733
Immunogen Recombinant human TUBB2b (1-445aa) purified from E. coli
Purification Protein G purified
Storage Buffer 50mM Sodium Borate
Storage Temperature Store at 4C in the dark.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty