Anti-beta II Tubulin B Mouse Monoclonal Antibody (DyLight 350) [clone: 5B3]

NBP1-74029UV
103402-786EA 536.92 USD
103402-786
Anti-beta II Tubulin B Mouse Monoclonal Antibody (DyLight 350) [clone: 5B3]
The beta II Tubulin B Antibody (5B3) [DyLight 350] from Novus Biologicals is a mouse monoclonal antibody to beta II Tubulin B. This antibody reacts with human, mouse. The beta II Tubulin B Antibody (5B3) [DyLight 350] has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: beta II Tubulin B
Clonality: Monoclonal
Clone: 5B3
Conjugation: DyLight 350
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR