Anti-beta II Tubulin B Mouse Monoclonal Antibody [clone: 6D11]

Supplier: Novus Biologicals
NBP2-46250
103404-970EA 663.82 USD
103404-970
Anti-beta II Tubulin B Mouse Monoclonal Antibody [clone: 6D11]
Antibodies
The beta II Tubulin B Antibody (6D11) from Novus Biologicals is a mouse monoclonal antibody to beta II Tubulin B. This antibody reacts with human, mouse, canine, monkey. The beta II Tubulin B Antibody (6D11) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunofluorescence.

Type: Primary
Antigen: beta II Tubulin B
Clonality: Monoclonal
Clone: 6D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Canine, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR