Anti-beta II Tubulin A Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: TU-12]

NB500-585AF488
103402-722EA 493.85 USD
103402-722
Anti-beta II Tubulin A Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: TU-12]
The beta II Tubulin A Antibody (TU-12) [Alexa Fluor« 488] from Novus Biologicals is a mouse monoclonal antibody to beta II Tubulin A. This antibody reacts with mammal. The beta II Tubulin A Antibody (TU-12) [Alexa Fluor« 488] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: beta II Tubulin A
Clonality: Monoclonal
Clone: TU-12
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Mammal
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR