Anti-beta II Tubulin A Mouse Monoclonal Antibody (DyLight 680) [clone: TU-13]

NB110-55384FR
103402-756EA 493.85 USD
103402-756
Anti-beta II Tubulin A Mouse Monoclonal Antibody (DyLight 680) [clone: TU-13]
The beta II Tubulin A Antibody (TU-13) [DyLight 680] from Novus Biologicals is a mouse monoclonal antibody to beta II Tubulin A. This antibody reacts with human, mouse, porcine, plant. The beta II Tubulin A Antibody (TU-13) [DyLight 680] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen.

Type: Primary
Antigen: beta II Tubulin A
Clonality: Monoclonal
Clone: TU-13
Conjugation: DyLight 680
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Porcine, Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR