Anti-beta II Tubulin A Mouse Monoclonal Antibody [clone: 4C8]

H00007280-M09
103333-474EA 553.85 USD
103333-474
Anti-beta II Tubulin A Mouse Monoclonal Antibody [clone: 4C8]
The beta II Tubulin A Antibody (4C8) from Novus Biologicals is a mouse monoclonal antibody to beta II Tubulin A. This antibody reacts with human. The beta II Tubulin A Antibody (4C8) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta II Tubulin A
Clonality: Monoclonal
Clone: 4C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR