Anti-beta II Tubulin A Mouse Monoclonal Antibody [clone: 2B2]

Supplier: Novus Biologicals
H00007280-M03
103333-464EA 798.46 USD
103333-464
Anti-beta II Tubulin A Mouse Monoclonal Antibody [clone: 2B2]
Antibodies
The beta II Tubulin A Antibody (2B2) from Novus Biologicals is a mouse monoclonal antibody to beta II Tubulin A. This antibody reacts with human, mouse, rat. The beta II Tubulin A Antibody (2B2) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta II Tubulin A
Clonality: Monoclonal
Clone: 2B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR