Anti-Beta Ig-h3/TGFBI Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00007045-D01P
103333-056EA 429 USD
103333-056
Anti-Beta Ig-h3/TGFBI Rabbit Polyclonal Antibody
Antibodies
The Beta Ig-h3 / TGFBI Antibody from Novus Biologicals is a rabbit polyclonal antibody to Beta Ig-h3 / TGFBI. This antibody reacts with human. The Beta Ig-h3 / TGFBI Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Beta Ig-h3/TGFBI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR