Anti-beta I Tubulin Mouse Monoclonal Antibody [clone: 2A1A9]

Supplier: Novus Biologicals
NBP2-52577
103404-950EA 635.51 USD
103404-950
Anti-beta I Tubulin Mouse Monoclonal Antibody [clone: 2A1A9]
Antibodies
The beta I Tubulin Antibody (2A1A9) from Novus Biologicals is a mouse monoclonal antibody to beta I Tubulin. This antibody reacts with human, mouse, rat, monkey. The beta I Tubulin Antibody (2A1A9) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence, Immunocytochemistry, CyTOF-ready.

CyTOF-ready

Type: Primary
Antigen: beta I Tubulin
Clonality: Monoclonal
Clone: 2A1A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR