Anti-Beta Hydroxysteroid Dehydrogenase Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00003284-D01P
103328-562EA 844.62 USD
103328-562
Anti-Beta Hydroxysteroid Dehydrogenase Rabbit Polyclonal Antibody
Antibodies
The Beta Hydroxysteroid Dehydrogenase Antibody from Novus Biologicals is a rabbit polyclonal antibody to Beta Hydroxysteroid Dehydrogenase. This antibody reacts with human. The Beta Hydroxysteroid Dehydrogenase Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Beta Hydroxysteroid Dehydrogenase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR