Anti-beta Galactosidase Mouse Monoclonal Antibody (DyLight 405) [clone: DC3 10D6]

NB100-2694V
103404-784EA 404.62 USD
103404-784
Anti-beta Galactosidase Mouse Monoclonal Antibody (DyLight 405) [clone: DC3 10D6]
The beta Galactosidase Antibody (DC3 10D6) [DyLight 405] from Novus Biologicals is a mouse monoclonal antibody to beta Galactosidase. This antibody reacts with all species. The beta Galactosidase Antibody (DC3 10D6) [DyLight 405] has been validated for the following applications: ELISA.

Type: Primary
Antigen: beta Galactosidase
Clonality: Monoclonal
Clone: DC3 10D6
Conjugation: DyLight 405
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR