Anti-beta Galactosidase Mouse Monoclonal Antibody (DyLight 350) [clone: DC1 4C7]

Supplier: Novus Biologicals
NB100-2619UV
103369-194EA 614.15 USD
103369-194
Anti-beta Galactosidase Mouse Monoclonal Antibody (DyLight 350) [clone: DC1 4C7]
Antibodies
The beta Galactosidase Antibody (DC1 4C7) [DyLight 350] from Novus Biologicals is a mouse monoclonal antibody to beta Galactosidase. This antibody reacts with bacteria. The beta Galactosidase Antibody (DC1 4C7) [DyLight 350] has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: beta Galactosidase
Clonality: Monoclonal
Clone: DC1 4C7
Conjugation: DyLight 350
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR