Anti-beta COP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15579
102706-260EA 635.51 USD
102706-260
Anti-beta COP Rabbit Polyclonal Antibody
Antibodies
The beta COP Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta COP. This antibody reacts with human. The beta COP Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: beta COP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR