Anti-beta COP Mouse Monoclonal Antibody [clone: 3E10.]

Supplier: Novus Biologicals
H00001315-M08
103324-196EA 798.46 USD
103324-196
Anti-beta COP Mouse Monoclonal Antibody [clone: 3E10.]
Antibodies
The beta COP Antibody (3E10.) from Novus Biologicals is a mouse monoclonal antibody to beta COP. This antibody reacts with human. The beta COP Antibody (3E10.) has been validated for the following applications: ELISA.

Type: Primary
Antigen: beta COP
Clonality: Monoclonal
Clone: 3E10.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR