Anti-beta Casein Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: F20.14]

Supplier: Novus Biologicals
NB100-2720H
103404-762EA 614.15 USD
103404-762
Anti-beta Casein Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: F20.14]
Antibodies
The beta Casein Antibody (F20.14) [HRP] from Novus Biologicals is a mouse monoclonal antibody to beta Casein. This antibody reacts with human. The beta Casein Antibody (F20.14) [HRP] has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.

Type: Primary
Antigen: beta Casein
Clonality: Monoclonal
Clone: F20.14
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR