Anti-beta B1 Crystallin Mouse Monoclonal Antibody [clone: 3D9]

Supplier: Novus Biologicals
H00001414-M03
103324-390EA 798.46 USD
103324-390
Anti-beta B1 Crystallin Mouse Monoclonal Antibody [clone: 3D9]
Antibodies
The beta B1 Crystallin Antibody (3D9) from Novus Biologicals is a mouse monoclonal antibody to beta B1 Crystallin. This antibody reacts with human. The beta B1 Crystallin Antibody (3D9) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: beta B1 Crystallin
Clonality: Monoclonal
Clone: 3D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR